Sodium Spoon
Sodium spoon
Catalog # 470343-578
Supplier: Wards
Fournisseur #: CH0737
Jump to:
Product Details & Documents
The medium-sized sodium spoon is specifically designed for accurate and controlled dispensing of sodium, ensuring safe and efficient handling in experiments.
- This spoon is for holding sodium under water
- By compressing handles the sodium spoon opens to allow sodium to be placed in the caged spoon
- Comprised of rust-proof wire stem and strong cage to safely enclose the sodium
- Cage measures 15 mm (0.59") diameter
- Over all Length 248 mm (9.75")
Spécifications
- Désignation:Sodium spoon