48-Peg Wall Mounted Draining Rack
48-peg wall mounted draining rack
Catalog # 470345-972
Supplier: Wards
Supplier #: CH0715D
Jump to:
Product Details & Documents
Made of vinyl-coated steel, this wall-mountable rack with 48 pegs (2.75" pegs) offers ample space for drying various labware with added durability.
- Test tube draining rack featuring a sturdy vinyl-coated steel construction that will stand up to the wear and tear of the lab
- Supports up to 36 test tubes with 2.75" pegs - Provides convenient insertion and removal of tubes
- Stands upright or hangs on the wall (mounting hardware not included)
- Designed for use in professional laboratories, but is an ideal choice for science classrooms
This sturdy, 20×20" wall mounted laboratory drying rack contains 48 test tube pins. Each pin is 2.75" long and pegs are 3" apart. The rack is vinyl coated steel and each peg features a rubber cap to prevent damage and scratches to glassware. Mounting Accessories not included.
Specifications
- Désignation:48-peg wall mounted draining rack