32-Peg Draining Rack
32-peg draining rack
Catalog # 470345-712
Supplier: Wards
Supplier #: CH0715B
Jump to:
Product Details & Documents
This metal rack features 32 pegs for efficient drying of labware, ensuring proper airflow and organization in the laboratory.
- Test tube draining rack featuring a sturdy vinyl-coated steel construction that will stand up to the wear and tear of the lab
- Supports up to 32 test tubes with 4.25" pegs - Provides convenient insertion and removal of tubes. 21.5" tall, 17.25" wide, 12" depth from the peg tips, and weighs 2.6 lbs.
- Stands upright or hangs on the wall (mounting hardware not included)
- Designed for use in professional laboratories, but is an ideal choice for science classrooms
Vinyl coated metal build is easy on glassware while being durable at the same time. Pegs are spaced 2.25" apart horizontally and 6/5.5/6.5" vertically to hold a variety of shapes and sizes; test tubes, beakers, flasks and more.
Spécifications
- Désignation:32-peg draining rack