2-Prong Adjustable Clamp
2-prong adjustable clamp
Catalog # 470344-376
Supplier: Wards
Fournisseur #: CH0680A
Jump to:
Product Details & Documents
Adjustable 2-prong clamp with an integral boss head, vinyl-coated non-slip jaws, and the ability to hold objects ranging from 8 to 30 mm in diameter, providing a versatile and secure gripping solution for various lab applications.
- Two prong, adjustable clamp
- Integral bosshead
- Universal- clamp is ideal for use with straight sided glassware and apparatus such as burettes, tubes and flask necks
- Jaws are coated with nonslip vinyl
- Clamp measures 8" in length
Spécifications
- Désignation:2-prong adjustable clamp