Universal Clamp
Universal clamp
Catalog # 470344-048
Supplier: Wards
: CH0679
Jump to:
Product Details & Documents
Universal clamp with 2 prongs and cork-lined jaws, providing a gentle yet secure grip for holding objects up to 2.3" wide.
- Adjustable jaws hold objects up to 60 mm in width
- Fitted with 15 cm long steel rod
- Cork lined jaws
- Perfect for test tubes, burettes, or flasks
Two prong, diecasted with other parts of plated brass, accepts articles from 15 to 60 mm diameter actuated by nickel plated brass pummels and twin screw.
Specifications
- Désignation:Universal clamp