Crystal Lattice Fullerene Model Kit
Catalog # 470344-004
Supplier: Wards
Specifications
- Description:Crystal lattice fullerene model kit
- Cat. no.:470344-004
Specifications
About this item
Crystal lattice fullerene model kit for educational purposes, providing hands-on exploration of molecular structures and properties.
- Includes: 60 black spheres and 90 medium connectors
- Black spheres represent atoms
- Gray connectors represent bonds between atoms
The crystal lattice fullerene model kit provides an ideal tool for studying the structure of a fullerene molecule in a visually and kinesthetically effective manner.