Platinum Wire | Avantor
Platinum Wire
Platinum wire
Catalog # 470344-750
Supplier:   Wards
Fournisseur #:  CH0538
Platinum Wire
Catalog # 470344-750
Supplier:  Wards
Platinum Wire
Platinum Wire
Platinum wire
Price
$23.85
Log in pour la disponibilité
Each
Product Details & Documents
Platinum wire supplied in a glass tube, designed specifically for flame tests, ensuring accurate and consistent results.
  • Platinum wire ideal for flame test
  • Supplied in borosilicate glass tube, fused into glass handle
  • Attached to silicon cork
  • Tube measures 5" long and 0.6" in diameter. Wire measures 18 to 20 mm long and approximately 0.12 mm in diameter
Spécifications
  • Désignation:
    Platinum wire