Single Burette Clamp
Single burette clamp
Catalog # 470343-856
Supplier: Wards
Fournisseur #: CH0256A
Jump to:
Product Details & Documents
Single burette clamp made of casted material with a built-in boss head.
- Casted, with built-in boss head
- Approximately 3.75" tall, 4.5" wide and 2.75" long
- To secure single burette
- Durable, non slip
- Basic essential for classroom and laboratories
Fits burette up to 25 mm.
Spécifications
- Désignation:Single burette clamp