Vacuum Distillation Set
Vacuum distillation set
Catalog # 470343-678
Supplier: Wards
Fournisseur #: CH1028
Jump to:
Product Details & Documents
5-piece set with 24/29 joints for precise vacuum distillation, made of durable borosilicate glass.
- Vacuum Distillation set made of high quality borosilicate glass
- Set includes 5 pieces: 250 ml round bottom flask, still head, Liebig condenser, adaptor receiver vacuum, 100 ml Erlenmeyer flask
- All joint sizes are 24/29
- All pieces are made of high quality, heavy duty borosilicate glass 3.3
- Perfect for any school or professional laboratory
Spécifications
- Désignation:Vacuum distillation set