Hydro Distillation Kit
Hydro distillation kit
Catalog # 470345-528
Supplier: Wards
Fournisseur #: CH0890C
Jump to:
Product Details & Documents
Designed for hydro distillation, this 9-piece kit includes a 250 ml flask and various components with 19/26 joints, offering convenience and reliability for extracting essential oils and other hydro-distilled substances in laboratory work.
- Distillation is the process of separating the component or substances from a liquid mixture by selective evaporation and condensation
- Kit includes 9 pieces :1 to 250 ml round bottom boiling flask two necks, 1 - still head plain, 1 - liebig condenser, 1 - receiver adapter delivery, 1 - dropping funnel 100 ml with stopper, and 4 joint clips 19/26
- All joints sizes are 19/26 on pieces
- All pieces are made of high quality borosilicate glass 3.3
- Clamps, stand, thermometer or bunsen burner are not included with this kit
Steam (Hydro) distillation is a special type of distillation for temperature sensitive materials, like aromatic compounds. This process of separating a component or substances from a liquid mixture by selective evaporation and condensation. Eisco Borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.
Spécifications
- Désignation:Hydro distillation kit