28-Peg Wall Mounted Draining Rack
28-peg wall mounted draining rack
470344-020
: Wards
: CH0715C
Designed for space-saving, this wall-mounted rack with 28 pegs provides an organized drying solution for laboratory equipment.
- Test tube draining rack featuring a sturdy vinyl-coated steel construction that will stand up to the wear and tear of the lab
- Supports up to 28 test tubes with 70 mm pegs - Provides convenient insertion and removal of tubes. Size 16×12" with 2 rows with 5 pegs, 3 rows with 6 pegs
- Stands upright or hangs on the wall (mounting hardware not included)
- Designed for use in professional laboratories, but is an ideal choice for science classrooms
The sturdy wall mounted drying rack contains 28 pegs (two rows of five pegs and three rows of six pegs). The rack is vinyl coated.
- Désignation:28-peg wall mounted draining rack