Plastic Single Burette Clamp
Plastic single burette clamp
Catalog # 470345-314
Supplier: Wards
Fournisseur #: CH0262A
Jump to:
Product Details & Documents
Single polypropylene burette clamp.
- Single clamp - fits rods 10 to 15 mm in diameter
- Features two tapered wedges and a spring-loaded jack
- Made of high quality polypropylene
The sturdy, polypropylene single burette clamp is a great tool for tightly holding 10 and 12.5 mm rods. The spring-loaded jack and two tapered wedges assure a snug and safe fit.
Spécifications
- Désignation:Plastic single burette clamp