Wearable Circuits
Wearable Circuits
Supplier:   BROWN DOG GADGETS
Wearable Circuits
Supplier:  BROWN DOG GADGETS
Certificates
Wearable Circuits
2 Options Available Below
Product Details & Documents
Introduce core science and engineering concepts within an arts context.
  • Engage makers of all ages
  • Put the art in STEAM activities - learn about circuits while crafting
  • No additional tools needed, not recommended for sewing machines
Engage learners of all ages by adding circuity to hand-crafted wearable items. Wearable Circuits kits make this idea approachable for all learners regardless of dexterity issues, classroom time constraints, or existing sewing skills. Use our conductive Maker Tape™ and raw circuit components to add a variety of LED accents to virtually any wearable item that you can stick a piece of tape to.
Not recommended for sewing machines. Fabric not included.
Delivery information: Standard pack includes (15) Jumbo LEDs, (5) CR2032 batteries, (5) Adhesive hook and loop dots, (1) 2" Conductive hook and loop adhesive strip, (2) Rolls of conductive Maker tape™, ¼" × 5 m, Downloadable instruction manual. Classroom pack includes (75) Jumbo LEDs; (25) CR2032 batteries; (25) Adhesive hook and loop dots, (1) 8" Conductive hook and loop adhesive strip; (2) Rolls of conductive Maker Tape™, ¼" × 20 m; Downloadable instruction manual.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Wearable circuits, standard pack
    Log in for availability
    Log in for availability
    Each
    Each - 30,10 $
    30,10 $
    Clearance
    Each
       30,10 $
    Supplier Number: K1060-STA
  • Wearable circuits, classroom pack
    Log in for availability
    Log in for availability
    Each
    Each - 181,99 $
    181,99 $
    Each
       181,99 $
    Supplier Number: K1060-CLS
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Conductive Maker Tape™, ¼" × 20 m
    Conductive Maker Tape™, ¼" × 20 m
    470234-666
    Log in for availability
    Each - $16.45