Synthware Springs
Supplier: Kemtech America
Jump to:
À propos de cet article
Stainless steel retention springs.
- Chemically resistant stainless steel
- Compatible with other brands
Stainless steel springs for use with glassware that features hooks.
2 Options Available Below
Product Details & Documents
Stainless steel retention springs.
- Chemically resistant stainless steel
- Compatible with other brands
Stainless steel springs for use with glassware that features hooks.