Synthware Plug, High Vacuum, Teflon® Protected, Metering
Supplier:   Kemtech America
Synthware Plug, High Vacuum, Teflon® Protected, Metering
Supplier:  Kemtech America
Certificates
Synthware Plug, High Vacuum, Teflon® Protected, Metering
Synthware Plug, High Vacuum, Teflon® Protected, Metering
2 Options Available Below
Product Details & Documents
Designed to provide an excellent vacuum tight seal without exposing any material other than Teflon® to chemicals.
  • Teflon shaft
  • PTFE covered Viton O-rings
Complete with control knob. Viton® O-rings on PTFE shaft are protected with two precision Teflon® wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal.
Product Family Options
Informations sur le produit
  • Désignation
    Stopcock Size
    Availability
    Price
  • High vacuum plug
    0 - 8 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $105.21
    $105.21
    Prix promotionnel pour les nouveaux clients
    Each
       $105.21
    Supplier Number: S660008
  • High vacuum plug
    0 - 12 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $131.99
    $131.99
    Prix promotionnel pour les nouveaux clients
    Each
       $131.99
    Supplier Number: S660012