Synthware Storage Flask, #15 O-Ring Joint
1000 ml, Storage flask, #15 O-ring joint, 220 m high, 0 - 8 mm
Catalog # 60004-700
Supplier: Kemtech America
Supplier #: S521000
Jump to:
Product Details & Documents
Storage Flask features a #15 O-ring joint on the sidearm and a high vacuum valve to control gas and/or liquid flow into the vessel.
- Heavy wall construction
- 33 expansion borosilicate glass
- Standard ground glass joints work with other brands
Supplied complete with one #116 Viton® O-ring, and one high vacuum valve.
Specifications
- Capacité:1000 ml
- Désignation:Storage flask, #15 O-ring joint, 220 m high
- Valve Size:0 - 8 mm