Synthware Replacement T-Bore Glass Stopcocks
Supplier: Kemtech America
Jump to:
À propos de cet article
Glass plug ONLY.
- Heavy wall construction
- 33 expansion borosilicate glass
- Standard ground glass joints work with other brands
For use with Kemtech S69 series stainless steel retention clips.
3 Options Available Below
Product Details & Documents
Glass plug ONLY.
- Heavy wall construction
- 33 expansion borosilicate glass
- Standard ground glass joints work with other brands
For use with Kemtech S69 series stainless steel retention clips.
Product Family Options
Informations sur le produit
- DésignationBore SizeAvailabilityPrice
- Supplier Number: S510217
- Supplier Number: S511740
- Supplier Number: S512044