Synthware Storage Vessel, #15 O-Ring Joint
Supplier:   Kemtech America
Synthware Storage Vessel, #15 O-Ring Joint
Supplier:  Kemtech America
Certificates
Synthware Storage Vessel, #15 O-Ring Joint
Synthware Storage Vessel, #15 O-Ring Joint
3 Options Available Below
Product Details & Documents
Storage vessel with a #15 O-ring joint on the sidearm and a high vacuum valve to control gas or liquid flow into the vessel.
  • Heavy wall construction
  • 33 expansion borosilicate glass
  • Standard ground glass joints work with other brands
Supplied complete with one #116 Viton® O-ring, and one high vacuum valve.
Product Family Options
Informations sur le produit
  • Capacité
    Désignation
    Valve Size
    Availability
    Price
  • 50 ml
    Storage vessel, #15 O-ring joint, 190 mm high
    0 - 4 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $238.56
    $238.56
    Prix promotionnel pour les nouveaux clients
    Each
       $238.56
    Supplier Number: S490050
  • 100 ml
    Storage vessel, #15 O-ring joint, 235 mm high
    0 - 8 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $269.90
    $269.90
    Prix promotionnel pour les nouveaux clients
    Each
       $269.90
    Supplier Number: S490100
  • 350 ml
    Storage vessel, #15 O-ring joint, 240 mm high
    0 - 8 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $301.73
    $301.73
    Prix promotionnel pour les nouveaux clients
    Each
       $301.73
    Supplier Number: S490350