Synthware #35 Ball Stopper
Ball stopper, #35
60004-858
: S450035
35/20 ball joint penny head stopper.
- Heavy wall construction
- 33 expansion borosilicate glass
- Standard ground glass joints work with other brands
For use with 35/20 socket joint.
- Désignation:Ball stopper
- Taille:#35