Synthware Check Valve
Check valve, 8 mm hose connection
Catalog # 60004-922
Supplier: Kemtech America
Fournisseur #: V180008
Jump to:
Product Details & Documents
Gound seat for liquid seal.
- Heavy wall borosilicate construction
- All glass
- Ground seat to ensure liquid seal
Heavy wall check valve is designed to prevent gas or liquid backflow.
Spécifications
- Type de connection:8 mm hose connection
- Désignation:Check valve