Certified Weight Set, Sartorius
Special weight set, 2×50 g, aluminium, for ME/MC with 0.01 mg or 0.1 mg, with certificate
Catalog # CA76413-052
Supplier:   Sartorius
 :  YSS45-00
Certified Weight Set, Sartorius
Catalog # CA76413-052
Supplier:  Sartorius
Certified Weight Set, Sartorius
Certified Weight Set, Sartorius
Special weight set, 2×50 g, aluminium, for ME/MC with 0.01 mg or 0.1 mg, with certificate
Price
$489.38
Log in pour la disponibilité
Each
Product Details & Documents
Special aluminimum weight set including two 50 g weights. Factory-certified.
Suitable for air density determinations.
Spécifications
  • Désignation:
    Special weight set, 2×50 g, aluminium, for ME/MC with 0.01 mg or 0.1 mg, with certificate