Certified Weight Set, Sartorius
Special weight set, 2×50 g, aluminium, for ME/MC with 0.01 mg or 0.1 mg, with certificate
Catalog # CA76413-052
Supplier: Sartorius
: YSS45-00
Jump to:
Product Details & Documents
Special aluminimum weight set including two 50 g weights. Factory-certified.
Suitable for air density determinations.
Spécifications
- Désignation:Special weight set, 2×50 g, aluminium, for ME/MC with 0.01 mg or 0.1 mg, with certificate