Cryogenic Storage Labels
 :   Computype
Cryogenic Storage Labels
 :  Computype
Cryogenic Storage Labels
Cryogenic Storage Labels
Wrap-around labels for glass or plastic tubes and vials exposed to temperatures as low as −196 °C.
  • TS1113 polypropylene
  • TR618Z ribbon: 52×153 mm
  • Self-adherese and completely wraps around tubes ranging in size from 0.5 to 50 ml
  • Withstands both direct and vapor liquid nitrogen exposure and an autoclave
  • Resistant to solvents such as DMSO, methanol, isopropyl alcohol and more
Designed to protect both print and adhesion, these labels wrap completely around tubes of ranging sizes and self-adhere. The self-laminating design protects sample integrity through cryogenic storage and various other common lab exposures. They can be applied to frozen glass, though it is recommended to wipe frost from the surface before application to ensure adhesion.
Both the 1 and 3'' core are compatible with thermal transfer printers. The 3'' core is compatible with the cab AXON2 and cab AXON1 printer/applicators.
 : Kit includes labels, print ribbon, and cleaning cloth.
Product Family Options
'' core Printers
  • Désignation
    Matière
    Dimensions
    Packaging
  • Wrap-around cryogenic liquid nitrogen label, with 0.5×0.75'' white patch
    TS1113 polypropylene
    0.5×1.8''
    1400 labels/kit
    Kit of 1
    Kit of 1 - $802.22
    $802.22
    Kit of 1
       $802.22
     : 1061485
  • Wrap-around cryogenic liquid nitrogen label, with 0.68×1'' white patch
    TS1113 polypropylene
    0.68×2.5''
    1000 labels/kit
    Kit of 1
    Kit of 1 - $821.42
    $821.42
    Kit of 1
       $821.42
     : 1061488
  • Wrap-around cryogenic liquid nitrogen label, with 1×1'' white patch
    TS1113 polypropylene
    1×2.625''
    950 labels/kit
    Kit of 1
    Kit of 1 - $714.02
    $714.02
    Kit of 1
       $714.02
     : 1061491
3'' core Printers
  • Désignation
    Matière
    Dimensions
    Packaging
  • Wrap-around cryogenic liquid nitrogen label, with 0.5×0.75'' white patch
    TS1113 polypropylene
    0.5×1.8''
    3000 labels/kit
    Kit of 1
    Kit of 1 - $1,063.72
    $1,063.72
    Kit of 1
       $1,063.72
     : 1061484
  • Wrap-around cryogenic liquid nitrogen label, with 0.68×1'' white patch
    TS1113 polypropylene
    0.68×2.5''
    2000 labels/kit
    Kit of 1
    Kit of 1 - $1,275.33
    $1,275.33
    Kit of 1
       $1,275.33
     : 1061487
  • Wrap-around cryogenic liquid nitrogen label, with 1×1'' white patch
    TS1113 polypropylene
    1×2.625''
    2000 labels/kit
    Kit of 1
    Kit of 1 - $856.58
    $856.58
    Kit of 1
       $856.58
     : 1061490