Dispenser Dropping Bottles
Supplier: United Scientific Supplies
Jump to:
À propos de cet article
Precision dispensing of solutions.
- Dispenses one drop at a time
- Spout lid is connected to the cap
- LDPE with leakproof PP cap
- BPA-free
This LDPE dispensing bottle can be easily operated with one hand. The bottle features a polypropylene screw cap with a tethered spout lid that snaps securely in place.
5 Options Available Below
Product Details & Documents
Precision dispensing of solutions.
- Dispenses one drop at a time
- Spout lid is connected to the cap
- LDPE with leakproof PP cap
- BPA-free
This LDPE dispensing bottle can be easily operated with one hand. The bottle features a polypropylene screw cap with a tethered spout lid that snaps securely in place.
Product Family Options
Informations sur le produit
- DésignationCapacitéMatièreAvailabilityPrice
- Supplier Number: UNDDB015
- Supplier Number: UNDDB030
- Supplier Number: UNDDB060
- Supplier Number: UNDDB125
- Supplier Number: UNDDB250