ContactUsLinkComponent
Light Measurement Kit
  470350-120
undefined
Light Measurement Kit
  470350-120
 :  654-0100
  • Description:
    Light measurement kit
  • Cat. no.:
    470350-120
Students will calculate the relationship between the apparent brightness of a luminous object and the distance to the object.
  • Hand-on-learning activities
    Include a light sensor
    Covers topics on light sources and optics
This Light Measurement Kit is designed to measure the intensity of light in various settings, making it a perfect tool for science experiments in classrooms.
 : The kit contains 10× paraffin blocks; 10 foot-long pieces of optical fiber; 1×200 W lamp; 1× lampholder; 20× rubber bands; 1× flashlight with batteries; 1× 25' roll of aluminium foil; 1 set of instructions and activities.