Cardboard Spectrometer Kit
About this item
Investigate light spectra with this easy-to-use spectrometer.
Recommended for astronomy, chemistry, physics, physical science and earth science
Easy to use
Offers a clear view of the light’s properties
This spectrometer is designed for educational purposes, helping students explore and understand light spectra. Observe absorption lines in the Sun's spectrum, the spectra of energized elements in gaseous states and the double yellow lines of Mercury vapors in the spectrum of a fluorescent lamp.
1 Options Available Below
Product Details & Documents
Investigate light spectra with this easy-to-use spectrometer.
Recommended for astronomy, chemistry, physics, physical science and earth science
Easy to use
Offers a clear view of the light’s properties
This spectrometer is designed for educational purposes, helping students explore and understand light spectra. Observe absorption lines in the Sun's spectrum, the spectra of energized elements in gaseous states and the double yellow lines of Mercury vapors in the spectrum of a fluorescent lamp.
Product Family Options
Product Information
- DésignationGradeAvailabilityPrice
- Supplier Number: 654-0025
- Cardboard spectrometer kit, pack of 10Discontinued-Supplier Number: -