Dropping Bottle
Supplier: United Scientific Supplies
Jump to:
À propos de cet article
Dispense solutions accurately, one drop at a time.
- Ideal for storing and dispensing small volumes
- Translucent design
- LDPE with leakproof PP cap
- BPA-free
Dropping bottles are useful for adding small amounts of a liquid. With a small squeeze the amount of liquid being dispensed can be controlled. Manufactured from translucent and low density polyethylene. Caps are made from polypropylene. Assembled bottles are sold as packs.
6 Options Available Below
Product Details & Documents
Dispense solutions accurately, one drop at a time.
- Ideal for storing and dispensing small volumes
- Translucent design
- LDPE with leakproof PP cap
- BPA-free
Dropping bottles are useful for adding small amounts of a liquid. With a small squeeze the amount of liquid being dispensed can be controlled. Manufactured from translucent and low density polyethylene. Caps are made from polypropylene. Assembled bottles are sold as packs.
Product Family Options
Informations sur le produit
- CapacitéMatièreAvailabilityPrice
- Supplier Number: PDB007PK12
- Supplier Number: PDB010PK12
- Supplier Number: PDB015PK12
- Supplier Number: PDB030PK12
- Supplier Number: PDB060PK12
- Supplier Number: PDB125PK12