Slide Storage Boxes, Hinged, Plastic
Supplier: United Scientific Supplies
Jump to:
À propos de cet article
Store slides securely and safely.
- Engraved numbers
- Secure closing clasp
- Key card for easy reference
Sturdy two-piece molded slide boxes hold standard 3×1" microscope slides. The hinged top has a reference card for labeling the corresponding slide description. The numbers on the opposite side are engraved for easy reference. These boxes are made from high impact polystyrene.
3 Options Available Below
Product Details & Documents
Store slides securely and safely.
- Engraved numbers
- Secure closing clasp
- Key card for easy reference
Sturdy two-piece molded slide boxes hold standard 3×1" microscope slides. The hinged top has a reference card for labeling the corresponding slide description. The numbers on the opposite side are engraved for easy reference. These boxes are made from high impact polystyrene.
Product Family Options
Informations sur le produit
- DésignationMatièreCapacitéAvailabilityPrice
- Supplier Number: 62211Spécifications:
More
Specifications
Cat. No.470349-718 - Supplier Number: 62212Spécifications:
More
Specifications
Cat. No.470349-770 - Supplier Number: 62213Spécifications:
More
Specifications
Cat. No.470349-780