Exact Volume Sampling Systems (EVSS)
Exact Volume Sampling Systems (EVSS)
Exact Volume Sampling Systems (EVSS)
Certificates
Exact Volume Sampling Systems (EVSS)
Exact Volume Sampling Systems (EVSS)
42 Options Available Below
Product Details & Documents
Optimize sampling processes with low yield, high value therapies with the Exact Volume Sampling Systems (EVSS).
  • Minimize drug product loss with a dual filtered air syringe system which enables precise sample collection as well as preservation of inline product volume
  • Sample outside of a bio-safety cabinet using a closed system assembly
Select a preset sampling volume ranging from 0.5 to 10.5 ml in 0.5 ml increments, ensuring consistent and accurate sampling of ±0.25 ml every time.
The EVSS is available with a TPE (C-Flex®) or PVC (Tygon®) inlet tubing option with a male luer end connection.
The EVSS can be aseptically connected to a single-use bioreactor or other source vessel using standard tube welders. It can also be aseptically sealed and removed (wet or dry) using tubing sealers such as the Genesis SE340 or handheld battery powered version SE640.
Accessories information: The single-use assemblies are individually double bagged, and gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶.
Documents
Product Family Options
TPE (C-Flex®) Inlet Tubing
Informations sur le produit
  • Capacité
    Matière
    Désignation
    Availability
    Price
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 0.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 0.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Case of 50
       $9,359.90
    Supplier Number: EVSS-CFLEX0.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 0.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 1.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 1.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX1.0
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 1.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 1.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 1.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX1.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 1.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 2.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 2.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX2.0
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 2.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 2.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 2.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX2.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 2.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 3.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 3.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX3.0
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 3.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 3.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 3.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX3.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 3.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 4.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 4.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX4.0
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 4.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 4.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 4.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX4.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 4.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 5.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 5.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX5.0
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 5.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 5.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 5.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX5.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 5.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 6.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 6.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX6.0
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 6.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 6.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 6.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX6.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 6.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 7.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 7.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX7.0
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 7.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 7.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 7.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX7.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 7.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 8.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 8.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX8.0
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 8.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 8.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 8.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX8.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 8.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 9.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 9.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX9.0
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 9.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 9.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 9.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX9.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 9.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 10.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 10.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX10.0
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 10.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 10.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 10.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,871.98
    $1,871.98
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,359.90
    $9,359.90
    Pack of 10
       $1,871.98
    Minimum order quantity is 1
    Case of 50
       $9,359.90
    Minimum order quantity is 1
    Supplier Number: EVSS-CFLEX10.5
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 10.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, C-Flex® inlet, 11.00 ml dip tube
    Discontinued
    Supplier Number: -
PVC (Tygon®) Inlet Tubing
Informations sur le produit
  • Capacité
    Matière
    Désignation
    Availability
    Price
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 8.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 8.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC8.0ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 0.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 0.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC0.5ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 0.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 1.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 1.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC1.0ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 1.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 1.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 1.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC1.5ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 1.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 2.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 2.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC2.0ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 2.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 2.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 2.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC2.5ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 2.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 3.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 3.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC3.0ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 3.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 3.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 3.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC3.5ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 3.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 4.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 4.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC4.0ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 4.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 4.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 4.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC4.5ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 4.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 5.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 5.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC5.0ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 5.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 5.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 5.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC5.5ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 5.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 6.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 6.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC6.0ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 6.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 6.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 6.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC6.5ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 6.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 7.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 7.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC7.0ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 7.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 7.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 7.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC7.5ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 7.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 8.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 8.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 8.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC8.5ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 8.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 9.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 9.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC9.0ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 9.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 9.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 9.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC9.5ML
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 9.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 10.00 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 10.00 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC10.0
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 10.25 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 10.50 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 10.50 ml dip tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $1,830.29
    $1,830.29
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $9,151.45
    $9,151.45
    Pack of 10
       $1,830.29
    Minimum order quantity is 1
    Case of 50
       $9,151.45
    Minimum order quantity is 1
    Supplier Number: EVSS-PVC10.5
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 10.75 ml dip tube
    Discontinued
    Supplier Number: -
  • 15 ml
    PP
    EVSS single-use assembly, Tygon® inlet, 11.00 ml dip tube
    Discontinued
    Supplier Number: -