Oligo Pro II System Capillary Arrays
Three different capillary arrays for analysis on the Oligo Pro II system.
- High resolution - optimized length allowing for n-1 resolution through 60 nt
- Variable throughput - 12, 24, and 96 capillary arrays designed to meet individual throughput demands
- Designed for the Oligo Pro II system - compatible with UV detection allowing for visualization of unlabeled oligonucleotides
The Oligo Pro II system has 3 capillary arrays, the 12-, 24-, and 96-Capillary Array Long, for analysis with the Oligo Pro Reagents. All three have a separation or effective length of 55 cm, with a total length of 80 cm. The 12 parallel capillary array is ideal for lower throughput, while the 24 and 96 are designed for medium to high-throughput laboratories.
: For research use only. Not for use in diagnostic procedures.
Frequently Bought Together