ZAG DNA Analyzer System
ZAG DNA analyzer system, High throughput capillary electrophoresis instrument for qualitative analysis of DNA fragments
  CA76645-238
ZAG DNA Analyzer System
  CA76645-238
ZAG DNA Analyzer System
ZAG DNA Analyzer System
ZAG DNA analyzer system, High throughput capillary electrophoresis instrument for qualitative analysis of DNA fragments
Each
Parallel capillary electrophoresis system for automated high-throughput DNA fragment analysis.
  • High separation resolution - reliably distinguish and size DNA fragments within 3 bp of each other
  • High throughput - process 48, 96-well sample trays in 24 hours. Broad sizing range - size fragments between 35 bp and 20,000 bp through a variety of available DNA analysis kits
  • Fast separation times - samples can be separated in as little as 20 minutes
  • Intuitive data analysis - analytical software can process separation data automatically after the run and generate a report in multiple formats
  • desktop PC supplied with a comprehensive 3-year warranty through HP
The Agilent ZAG DNA analyzer system relieves bottlenecks in automated high-throughput DNA fragment analysis workflows. Using parallel capillary electrophoresis, the ZAG DNA Analyzer system can process thousands of DNA fragment samples a day, eliminating bottlenecks associated with agarose gels.
This system is ideal for a range of DNA fragment applications, from basic PCR fragment analysis to microsatellite detection. Minimal sample preparation, automated separation of samples, and the option for continuous operation help keep your workflow moving.
 : For research use only. Not for use in diagnostic procedures.
  • Désignation:
    ZAG DNA analyzer system
  • Details:
    High throughput capillary electrophoresis instrument for qualitative analysis of DNA fragments
Accessories
  •  
    Description
    Details
  • CA76645-498
    96 well trays for buffer and waste locations, 50 per pack
    For use with the fragment analyzer, Femto Pulse, and ZAG DNA analyzer systems
    Pack of 1 - $844.00
  •  
    Description
    Details
  • CA76645-496
    Large open waste tray/tub
    For use with the fragment analyzer, Femto Pulse, and ZAG DNA analyzer systems
    Each - $43.80
  •  
    Description
    Details
  • CA76645-138
    dsDNA inlet buffer, 5X 930, 300 ml
    For use with the Fragment analyzer systems
    Each - $565.00
  •  
    Description
    Details
  • CA76645-254
    dsDNA inlet buffer 5X 930, 125 ml
    Running buffer for use with the Fragment analyzer systems
    Each - $194.01
  •  
    Description
    Details
  • CA76645-076
    Intercalating dye, 30 µl. Binding dye for nucleic acids, store at –20 ℃
    For use with the Fragment analyzer and ZAG DNA analyzer systems.
    Each - $232.00
  •  
    Description
    Details
  • CA76645-224
    Capillary conditioning solution, 5X, 100 ml
    Solution necessary for conditioning and restoring capillary array performance. For use with the Fragment analyzer and ZAG DNA analyzer systems
    Each - $185.00
  •  
    Description
    Details
  • CA76645-308
    Capillary conditioning solution, 5X, 50 ml
    Solution necessary for conditioning and restoring capillary array performance. For use with the Fragment analyzer and ZAG DNA analyzer systems
    Each - $92.40