TapeStation System, 4150 | Avantor
TapeStation System, 4150
4150 TapeStation System, Includes software, vortexer, accessories, plastic consumables, user information and I+F, does not include ScreenTape consumables and reagents
Catalog # CA76645-124
Fournisseur #:  G2992AA
TapeStation System, 4150
Catalog # CA76645-124
TapeStation System, 4150
TapeStation System, 4150
4150 TapeStation System, Includes software, vortexer, accessories, plastic consumables, user information and I+F, does not include ScreenTape consumables and reagents
Price
Renseignez-vous sur le prix
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Economic, entry-level system for the analysis of up to 16 DNA and RNA samples per run.
  • Established ScreenTape technology guarantees superior ease-of-use of the system, constant costs per sample for up to 16 samples per run
  • Wide range of RNA and DNA ScreenTape applications available including assays for genomic and cell-free DNA
  • Results are obtained quickly in as little as 1 to 2 minutes per sample, or less than 20 minutes for 16 samples
  • Easily switch between DNA and RNA ScreenTape assays for excellent application flexibility, Integrity standards for RNA (RNA integrity number equivalent, RINe) and genomic DNA (DNA Integrity Number, DIN) available
  • Compact footprint saves precious bench space in the laboratory, seamless upgrade to 4200 TapeStation possible by complete assay compatibility
The new agilent 4150 TapeStation system complements the agilent automated electrophoresis platforms with a new, compact, low-throughput instrument. Building on the success of the proven ScreenTape technology, it allows a quick, easy and reliable analysis of nucleic acids for up to 16 samples per run. This affordable, entry-level system is the ideal solution for the quality control (QC) of RNA and DNA samples in next-generation sequencing (NGS), microarray and qPCR workflows.
Caution: For research use only. Not for use in diagnostic procedures.
Spécifications
  • Désignation:
    4150 TapeStation System
  • Details:
    Includes software, vortexer, accessories, plastic consumables, user information and I+F, does not include ScreenTape consumables and reagents
Accessories
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Loading tips, 10 pack
    Loading tips, 10 pack
    CA76645-142
    For use with Agilent 4150 and 4200 TapeStation systems, 1 package contains 112 tips
    Log in pour la disponibilité
    Pack of 10 - $1,010.00
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • 32-pin electrode cartridge
    32-pin electrode cartridge
    CA76645-478
    For use with 4150 TapeStation (G2992A) and new 4200 TapeStation instruments (G2991B) only
    Log in pour la disponibilité
    Each - $677.00
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Loading tips, 1 pack
    Loading tips, 1 pack
    CA76645-170
    For use with Agilent 4150 and 4200 TapeStation systems, 1 package contains 112 tips
    Log in pour la disponibilité
    Pack of 1 - $96.80
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • USB cable, male A - male B
    USB cable, male A - male B
    CA76645-038
    For use with Agilent 4150 and 4200 TapeStation systems
    Log in pour la disponibilité
    Each - $75.20
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Needle change cartridge
    Needle change cartridge
    CA76645-314
    For use with Agilent 4150 and 4200 TapeStation systems
    Log in pour la disponibilité
    Each - $489.00