Bioanalyzer High Sensitivity DNA Analysis | Avantor
Bioanalyzer High Sensitivity DNA Analysis
Bioanalyzer High Sensitivity DNA Analysis
Certificates
Bioanalyzer High Sensitivity DNA Analysis
Bioanalyzer High Sensitivity DNA Analysis
2 Options Available Below
Product Details & Documents
Quality control of low input NGS libraries or DNA fragments up to 7000 bp.
  • Increased sensitivity for DNA analysis, down to 5 pg/µl for fragment analysis or 100 pg/µl for complex DNA samples, such as NGS libraries
  • A broad linear dynamic range that allows detection of less abundant PCR products
  • Accurate smear analysis within a specified size range of interest
  • Enhanced sizing accuracy, normalized to internal markers and external ladder
  • Quantitation accuracy and reproducibility calculated against precise internal standards
  • Minimal sample consumption is required as only 1 µl of sample is needed for analysis
  • Minimal exposure to hazardous materials, such as ethidium bromide, ensures safety
  • Achieve improved quality control analysis of next-generation sequencing (NGS) library smears derived from just a few amplification cycles
High Sensitivity DNA electrophoresis with the Bioanalyzer system enables improved quality control analysis of next-generation sequencing (NGS) library smears derived from just a few amplification cycles. Whether fragmented DNA or DNA sequencing libraries, the Bioanalyzer High Sensitivity DNA Kit reliably sizes and quantifies limited samples in the lower pg/µl concentration range.
The High Sensitivity DNA assays are often used for sample quality control for next-generation sequencing libraries. In addition, PCR amplicon analysis, multiplex PCR amplicons, gene expression analysis by RT-PCR, restriction fragment analysis, and detection of targeted cleavage in gene editing studies may all be carried out. Accuracy is ensured with a broad linear dynamic range and normalization to internal markers and ladder.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Details
    Availability
    Price
  • Agilent High sensitivity DNA kit
    For the separation, sizing and quantification of dsDNA samples of limited abundance and sized from 50 - 7000 bp, the kit includes 10 microfluidic chips, reagents and consumables
    Note: Incompatible with G2938A Bioanalyzer instruments
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,660.00
    $1,660.00
    Each
       $1,660.00
    La quantité minimale de commande est de 1
    Supplier Number: 5067-4626
  • Agilent High sensitivity DNA reagents
    Reagents only, no chips included, sufficient for 10 chips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $745.45
    $745.45
    Each
       $745.45
    La quantité minimale de commande est de 1
    Supplier Number: 5067-4627