These flammable storage refrigerators offer premium cooling and protection for flammable substances.
- Microprocessor temperature controller
- Temperature alarms
- Keyed door locks
- Probe access
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These flammable storage refrigerators meet the requirements of NFPA 45 and NFPA 30 for the cold storage of flammable and combustible materials in unclassified laboratory locations. They utilize non-sparking and/or ATEX rated components to ensure an intrinsically safe interior for flammable materials in sealed containers. These units feature temperature alarms, keyed door locks, interior shelving, probe access, and microprocessor temperature control.
The Plus Series includes premium additions such as extensive alarm systems and digital touch pad displays. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
*Select models ENERGY STAR certified: 76644-722
- Catalog No:
- 76644-726
- 76644-722
- 76644-718
- Model:
- 76644-726
- 76644-722
- 76644-718
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Defrost:
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Shelves/drawers:
- 5 adjustable shelves (4 full, 1 half shelf)
- 5 adjustable shelves (4 full, 1 half shelf)
- 5 adjustable shelves (4 full, 1 half shelf)
- Door type:
- 1 solid door, right hinged
- 1 solid door, right hinged
- 1 solid door, right hinged
- Weight:
- 114 kg (251 lbs.)
- 190 kg (241 lbs.)
- 87 kg (191 lbs.)
- Environmental certification:
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants