Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Pouring ring, MicroSolv

Pouring ring, MicroSolv

Replacement pouring ring or drip ring, PP, HPLC mobile phase reservoir bottles, Blue, GL 45
Catalog # 97056-780
Supplier #:  58802-30
Pouring ring, MicroSolv
Catalog # 97056-780
Pouring ring, MicroSolv
Pouring ring, MicroSolv

Replacement pouring ring or drip ring, PP, HPLC mobile phase reservoir bottles, Blue, GL 45

Price
$3.60
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Using this Polypropylene Pouring or 'Drip Ring' can make decanting liquids from any bottle with a GL 45 Thread easier and prevent that last drop from getting away from you.

Friction fit makes this ring stay in the bottle during pouring and capping.

Specifications
  • Color:
    Blue
  • Désignation:
    Replacement pouring ring or drip ring
  • Matière:
    PP
  • For Use With:
    HPLC mobile phase reservoir bottles
  • Epaisseur:
    GL 45
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.