HPLC Mobile Phase Storage Bottle, MicroSolv
Bottles, HPLC mobile phase storage, GL45, 100 ml
Catalog # 75834-186
Supplier: MicroSolv Technology Corp
Fournisseur #: 58807-100-A-CS
Jump to:
Product Details & Documents
These amber borosilicate glass bottles have a 100 ml 'Total Fill Capacity' and are perfect as HPLC Mobile Phase Storage Bottles. The small openings and enamel graduations and marking patches makes them a convenient way to store concentrated Mobile Phase or its Ingredients.
Blue, solid screw caps without liners and polypropylene drip rings are included. This bottle is not 'safety coated'.
Spécifications
- Capacité:100 ml
- Désignation:Bottles, HPLC mobile phase storage, GL45
Accessories
Informations sur le produit
- Catalog #MaterialFor Use WithAvailabilityPrice
- Replacement pouring ring or drip ringReplacement pouring ring or drip ring97056-780PPHPLC mobile phase reservoir bottlesLog in pour la disponibilitéEach - $3.60