VWR® Stackable Mini Rollers
VWR® Stackable Mini Rollers
Supplier:   VWR International
VWR® Stackable Mini Rollers
Supplier:  VWR International
Certificates
VWR® Stackable Mini Rollers
2 Options Available Below
Product Details & Documents
VWR® Stackable mini rollers, designed for gentle agitation of multiple sized bottles/tubes in a space saving footprint.
  • Adjustable digitial speed control, timer
  • Sealed electronics, constructed of corrosion resistant materials, suitable for use in CO₂/95% Humidity environments
  • Remote control (wired) included, to control unit without disrupting CO₂ Incubator chamber environment
  • 3D swing model (76670-460) offers 18 mm (0.71") swing motion
  • Easy to add and remove rollers to accommodate a wide variety of tube/bottle sizes
  • Optional stacking kit to stack 2 units, maximzing bench space
All models include O-rings for 50 ml tubes and 15 ml tubes for smooth rolling and wired remote control module.
Certifications: CE, CSA.
Delivery information: Supplied with 76670-454, 76670-452, 76670-450, 2 year warranty.
Specifications for Product Family
Specifications
  • Catalog No:
  • 76670-460
  • 76670-462
  • Max. capacity:
  • 6.5 kg (14.33 lbs.), 3×1500 ml bottles
  • 6.5 kg (14.33 lbs.), 3×1500 ml bottles
  • Speed range:
  • 0.5 - 80 rpm
  • 0.5 - 80 rpm
  • Dimensions:
  • 360×378×72 mm (14.17×14.88×2.83"); Roller (Ø×L): 28×269 mm (1.1×10.6")
  • 360×378×72 mm (14.17×14.88×2.83"); Roller (Ø×L): 28×269 mm (1.1×10.6")
Specifications
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Stackable mini roller, 3D swing motion
    Log in for availability
    Log in for availability
    Each
    Each - $2,460.88
    $2,460.88
    New Customer Promotional Price
    Each
       $2,460.88
    Specifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    76670-460
    Max. capacity
    6.5 kg (14.33 lbs.), 3×1500 ml bottles
    Speed range
    0.5 - 80 rpm
    Dimensions
    360×378×72 mm (14.17×14.88×2.83"); Roller (Ø×L): 28×269 mm (1.1×10.6")
    2 year warranty
  • Stackable mini roller
    Log in for availability
    Log in for availability
    Each
    Each - $2,381.49
    $2,381.49
    New Customer Promotional Price
    Each
       $2,381.49
    Specifications:
    2 year warranty
    More
    Specifications
    Cat. No.
    76670-462
    Max. capacity
    6.5 kg (14.33 lbs.), 3×1500 ml bottles
    Speed range
    0.5 - 80 rpm
    Dimensions
    360×378×72 mm (14.17×14.88×2.83"); Roller (Ø×L): 28×269 mm (1.1×10.6")
    2 year warranty
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Bars for stacking
    Bars for stacking
    76670-448
    Log in for availability
    Pack of 4 - $114.32
Product Information
  • Catalog #
    Availability
    Price
  • Control box
    Control box
    76670-450
    Log in for availability
    Each - $323.87
Product Information
  • Catalog #
    Availability
    Price
  • O-rings for 50 ml tube
    O-rings for 50 ml tube
    76670-454
    Log in for availability
    Pack of 20 - $9.52
Product Information
  • Catalog #
    Availability
    Price
  • O-rings for 15 ml tube
    O-rings for 15 ml tube
    76670-452
    Log in for availability
    Pack of 20 - $9.52
Product Information
  • Catalog #
    Availability
    Price
  • Rollers 2D
    Rollers 2D
    76670-456
    Log in for availability
    Pack of 2 - $76.21
Product Information
  • Catalog #
    Availability
    Price
  • Rollers 3D rocker
    Rollers 3D rocker
    76670-458
    Log in for availability
    Pack of 2 - $107.96