Backpack Lab Soil Quality Educational Test Kit
À propos de cet article
Backpack Lab designed for educators and environmental science students, containing lessons, activities, and test kits that relate to important parameters evaluated in soil quality.
- Test kits for the determination of common parameters in soil quality testing: nitrogen, phosphorus, potassium and pH.
Backpack Lab is designed with all the necessary components in one place, reducing the chance of misplacing an item. Ideal for transporting, it's easy to take this durable backpack to the field for on-site measurements. This portable kit contains well-constructed lessons and activities, and will allow the teacher to get the most out of their classroom time. The Combo pH/EC/TDS Tester is also included for measurement of pH, conductivity (EC), and total dissolved solids (TDS), as well as the T-Shaped Thermometer for accurate temperature measurements.
1 Options Available Below
Product Details & Documents
Backpack Lab designed for educators and environmental science students, containing lessons, activities, and test kits that relate to important parameters evaluated in soil quality.
- Test kits for the determination of common parameters in soil quality testing: nitrogen, phosphorus, potassium and pH.
Backpack Lab is designed with all the necessary components in one place, reducing the chance of misplacing an item. Ideal for transporting, it's easy to take this durable backpack to the field for on-site measurements. This portable kit contains well-constructed lessons and activities, and will allow the teacher to get the most out of their classroom time. The Combo pH/EC/TDS Tester is also included for measurement of pH, conductivity (EC), and total dissolved solids (TDS), as well as the T-Shaped Thermometer for accurate temperature measurements.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- 470017-358
- CA470017-358
- Accuracy:
- ±0.05 pH; ±2% F.S. (Conductivity); ±2% F.S. (TDS)
- ±0.05 pH; ±2% F.S. (Conductivity); ±2% F.S. (TDS)
- Calibration:
- Automatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or 4.01/6.86/9.18); automatic, one point at 1413 μS/cm or 1382 mg/L (ppm) (TDS)
- Automatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or 4.01/6.86/9.18); automatic, one point at 1413 μS/cm or 1382 mg/L (ppm) (TDS)
- Resolution:
- 0.01 pH; 1 μS/cm(Conductivity); 1 mg/L (ppm)(TDS)
- 0.01 pH; 1 μS/cm(Conductivity); 1 mg/L (ppm)(TDS)
Spécifications
Product Family Options
Informations sur le produit
- DésignationRangeAvailabilityPrice
- Backpack lab soil quality educational test kit0 - 14 pH; 0 - 3999 μS/cm (conductivity); 0 - 2000 mg/L (ppm) (TDS)Discontinued-Supplier Number: HI3896BPSpécifications:
More
Specifications
Cat. No.470017-358Accuracy±0.05 pH; ±2% F.S. (Conductivity); ±2% F.S. (TDS)CalibrationAutomatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or 4.01/6.86/9.18); automatic, one point at 1413 μS/cm or 1382 mg/L (ppm) (TDS)Resolution0.01 pH; 1 μS/cm(Conductivity); 1 mg/L (ppm)(TDS) - Backpack lab soil quality educational test kit0 - 14 pH; 0 - 3999 μS/cm (conductivity); 0 - 2000 mg/L (ppm) (TDS)Log in pour la disponibilitéEach$663.69Supplier Number: HI3896BPSpécifications:
More
Specifications
Cat. No.CA470017-358Accuracy±0.05 pH; ±2% F.S. (Conductivity); ±2% F.S. (TDS)CalibrationAutomatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or 4.01/6.86/9.18); automatic, one point at 1413 μS/cm or 1382 mg/L (ppm) (TDS)Resolution0.01 pH; 1 μS/cm(Conductivity); 1 mg/L (ppm)(TDS)