Backpack Lab Soil Quality Educational Test Kit | Avantor
Backpack Lab Soil Quality Educational Test Kit
Supplier:   Hanna
Backpack Lab Soil Quality Educational Test Kit
Supplier:  Hanna
Certificates
Backpack Lab Soil Quality Educational Test Kit
Backpack Lab Soil Quality Educational Test Kit
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Backpack Lab designed for educators and environmental science students, containing lessons, activities, and test kits that relate to important parameters evaluated in soil quality. 
  • Test kits for the determination of common parameters in soil quality testing: nitrogen, phosphorus, potassium and pH.
Backpack Lab is designed with all the necessary components in one place, reducing the chance of misplacing an item. Ideal for transporting, it's easy to take this durable backpack to the field for on-site measurements. This portable kit contains well-constructed lessons and activities, and will allow the teacher to get the most out of their classroom time. The Combo pH/EC/TDS Tester is also included for measurement of pH, conductivity (EC), and total dissolved solids (TDS), as well as the T-Shaped Thermometer for accurate temperature measurements.
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • 470017-358
  • CA470017-358
  • Accuracy:
  • ±0.05 pH; ±2% F.S. (Conductivity); ±2% F.S. (TDS)
  • ±0.05 pH; ±2% F.S. (Conductivity); ±2% F.S. (TDS)
  • Calibration:
  • Automatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or 4.01/6.86/9.18); automatic, one point at 1413 μS/cm or 1382 mg/L (ppm) (TDS)
  • Automatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or 4.01/6.86/9.18); automatic, one point at 1413 μS/cm or 1382 mg/L (ppm) (TDS)
  • Resolution:
  • 0.01 pH; 1 μS/cm(Conductivity); 1 mg/L (ppm)(TDS)
  • 0.01 pH; 1 μS/cm(Conductivity); 1 mg/L (ppm)(TDS)
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Range
    Availability
    Price
  • Backpack lab soil quality educational test kit
    0 - 14 pH; 0 - 3999 μS/cm (conductivity); 0 - 2000 mg/L (ppm) (TDS)
    Discontinued
    Supplier Number: HI3896BP
    Spécifications:
    More
    Specifications
    Cat. No.
    470017-358
    Accuracy
    ±0.05 pH; ±2% F.S. (Conductivity); ±2% F.S. (TDS)
    Calibration
    Automatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or 4.01/6.86/9.18); automatic, one point at 1413 μS/cm or 1382 mg/L (ppm) (TDS)
    Resolution
    0.01 pH; 1 μS/cm(Conductivity); 1 mg/L (ppm)(TDS)
  • Backpack lab soil quality educational test kit
    0 - 14 pH; 0 - 3999 μS/cm (conductivity); 0 - 2000 mg/L (ppm) (TDS)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $663.69
    $663.69
    Each
       $663.69
    Supplier Number: HI3896BP
    Spécifications:
    More
    Specifications
    Cat. No.
    CA470017-358
    Accuracy
    ±0.05 pH; ±2% F.S. (Conductivity); ±2% F.S. (TDS)
    Calibration
    Automatic, one or two-point with two sets of standard buffers (pH 4.01/7.01/10.01 or 4.01/6.86/9.18); automatic, one point at 1413 μS/cm or 1382 mg/L (ppm) (TDS)
    Resolution
    0.01 pH; 1 μS/cm(Conductivity); 1 mg/L (ppm)(TDS)