Simulaids® Alex Patient Simulators
À propos de cet article
Sees, listens, and responds.
- Three levels of options
- Iris cam for patient perspective
- One year warranty
The first patient communication simulator that sees, listens, and responds using artificial intelligence (AI). The Alex is an untethered, on-the-go patient simulator. Simply unpack, and you’re ready to train anytime and anywhere (Wi-Fi required). Speaks and understands English, French, and Spanish. Alex learns new languages all the time so it can help train more learners all around the globe.
Ordering information: These items have a limited return window. Please contact a sales rep if you want a preview or demonstration.
5 Options Available Below
Product Details & Documents
Sees, listens, and responds.
- Three levels of options
- Iris cam for patient perspective
- One year warranty
The first patient communication simulator that sees, listens, and responds using artificial intelligence (AI). The Alex is an untethered, on-the-go patient simulator. Simply unpack, and you’re ready to train anytime and anywhere (Wi-Fi required). Speaks and understands English, French, and Spanish. Alex learns new languages all the time so it can help train more learners all around the globe.
Ordering information: These items have a limited return window. Please contact a sales rep if you want a preview or demonstration.
Product Family Options
Lite
Informations sur le produit
Plus
Informations sur le produit
PRO
Informations sur le produit
- DésignationAvailabilityPrice
- Alex, light skinDiscontinued-Supplier Number: -
- Supplier Number: 101-7150M