Eisco® Chain Clamp
Chain clamp
470344-788
: Wards
: PRCMP21
Powder-coated die-cast Clamp with 19" long stainless steel chain, PVC-coated grip, and 5.5" long extention rod.
- Holds labware between 60 to 165 mm diameter securely in place at any angle to rods and lattices
- 19" long stainless steel chain loops around labware and tightens via threaded knob attachment to adjustable slide
- PVC coated fingers for slip resistant grip of beakers, bottles, and other labware
- Durable die-cast powder coated construction
Securely hold larger beakers, bottles, and other labware between 60 to 165mm in diameter with this versatile die-cast, powder-coated clamp. The attached 19" stainless steel chain loops around labware and hooks to a movable slide, holding it securely against the PVC-coated grip. A screw adjustment allows for precise pressure when gripping glass objects, to avoid potential breakage. A 5.5" long, 12 mm diamater extension rod can be attached to a support rod via a bosshead (available separately) and allows the clamp to be rotated 360°.
- Désignation:Chain clamp