ContactUsLinkComponent
SpectraMax® Mini Microplate Readers, Multi-Mode, Molecular Devices
undefined
SpectraMax® Mini Microplate Readers, Multi-Mode, Molecular Devices
  • Catalog No:
  • CA76640-490
  • CA76640-488
  • CA76640-508
  • CA76640-486
  • CA76640-506
  • CA76640-492
  • CA76640-496
  • CA76640-494
  • CA76640-504
  • CA76640-502
  • CA76640-500
  • CA76640-498
  • Temperature range:
  • 4 °C above ambient to 45 °C
  • 4 °C above ambient to 45 °C
  • 4 °C above ambient to 45 °C
  • W×D×H:
  • 400×320×350 mm
  • 400×320×350 mm
  • 400×320×350 mm
  • Weight:
  • 18 kg
  • 18 kg
  • 18 kg
Frequently Bought Together