Solar Motion Model
Solar Motion Model
Solar motion model
Catalog # 470349-334
Supplier:   Science First
Supplier #:  10240
Solar Motion Model
Catalog # 470349-334
Supplier:  Science First
Solar Motion Model
Solar Motion Model
Solar motion model
Price
$402.90
Log in for availability
Each
Product Details & Documents
Clearly shows the path of the sun at any latitude during any month of the year.
  • Elegant, easy to use, and hands-on model
  • Wooden desk top model with clean finish
  • Stands 13½" high
Ever wonder, 'What is the path of the sun where you live on Dec 31 or June 15?' This device will teach students how this astronomical feature is calculated.
Specifications
  • Désignation:
    Solar motion model