Hyclone™ Fetal Bovine Serum Alternative, Alpha Calf Fraction (United States), HyClone products (Cytiva)
Catalog # CA76635-856
Supplier: Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Serum format:Frozen
- Sterility:Sterile filtered
- Description:Alpha calf fraction, US origin, without iron
- Size:500 ml
- Cat. no.:CA76635-856
Specifications
About this item
HyClone™ Alpha Calf Fraction provides hybridoma users with a cost-effective medium supplement that promotes growth and antibody production.