Mag-Bind® PX blood RNA 96 kit, Omega Bio-Tek
Catalogue # CA76539-256
Fournisseur: Omega Bio-Tek
Numéro CAS:
Spécifications
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Blood DNA/RNA collection tubes (e.g. PAXgene )
- Target molecule:RNA
- Description:Mag-Bind® PX blood RNA kit (1×96 preps)
- Sample size:1× PAXgene® tube
- Cat. no.:CA76539-256
Spécifications
À propos de cet article
The Mag-Bind® PX blood RNA kit allows for the isolation of total RNA from 2,5 ml of whole blood that has been stored in PAXgene® Blood RNA tubes. Stabilised blood samples are first spun down and the nucleic acid pellets are washed and recollected. After washing, the nucleic acid pellet is dissolved into a specially formulated resuspension buffer. At this point the samples can be transferred to 96-well plates for processing in robotic liquid handlers and magnetic processors. High quality intact RNA can be isolated in less than 90 minutes.