SatPax® Stencil Cleaner 100% Polyester Nonwoven Cleanroom Wipes, Berkshire
Supplier: Berkshire
Certificates
About this item
Berkshire SatPax® Stencil Cleaner Wipes with Zestron Vigon® SC 200 is a powerful combination of low lint wiping material and effective stencil chemistry unmatched in the industry. SatPax® Stencil Cleaner Wipes are safe, fast-drying, and odor-free.
- Exceptional cleanliness
- Smooth and durable with good wet strength
- Non-shedding and low linting
- Knife cut, perforated roll
- Chemically compatible with common cleaning and disinfecting solutions
- Increases cleaning efficiency and cleaning protocol consistency
- No visible staining to stencil
Berkshire SatPax® Stencil Cleaner Wipes with Zestron Vigon® SC 200 quickly and effectively remove all types of solder pastes and adhesives used in electronic assembly manufacturing. These wipes are an ideal replacement for alcohol-based cleaners, which may not yield satisfactory cleaning results and are prohibited due to safety and environmental restrictions on VOC emissions or waste disposal issues.
Certifications: ISO Class 5 – 8. Cleanroom Class 100 – 100,000. RoHS I, II, and III, WEEE, and REACH SVHC.
Frequently Bought Together