ContactUsLinkComponent
Starter Pak No. 3, Gibco™ (a part of Thermo Fisher Scientific)
  CA76628-920
 :  
Starter Pak No. 3, Gibco™ (a part of Thermo Fisher Scientific)
  CA76628-920
 :  
  • Description:
    Starter Pak no. 3
  • Size:
    1 each
  • Cat. no.:
    CA76628-920
Starter Pak No. 3 provides a variety of yeast and soy products, ideal for processes where an animal origin-free medium is preferred. It provides conveniently packaged, 100 g, non-GMP samples of these peptones that are best suited for human and animal vaccine production.
Provides conveniently packaged, 100-gram, non-GMP samples of these peptones that are best suited for human and animal vaccine production and contains:
Bacto Yeast Extract and Gibco Yeast Extract - these peptones contain a mixture of peptides, amino acids, carbohydrates, and vitamins to support optimal growth in microbial species, and are ideal for the production of vaccines.
Bacto TC Yeastolate - this peptone is ideal for CHO-based applications of biotherapeutic monoclonal antibodies and recombinant proteins, as well as vaccine applications.
Phytone Peptone and Difco Soytone - these soy-based supplements provide a nutritious source of carbohydrates for successful use in microbial fermentation. Blending these soy peptones with yeast peptones has been shown to provide additional benefits to cultures.
Bacto Malt Extract - a water-soluble portion of malted barley, this peptone also provides carbohydrates for a variety of microbial fermentation processes.
The Starter Paks provide conveniently packaged samples of the peptones most commonly used in mammalian cell culture and microbial fermentation bioproduction processes. Starter Paks are tailored for specific applications, including the production of monoclonal antibodies, recombinant proteins, and vaccines.