Phytone™ Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Phytone Peptone is an animal origin-free (AOF) enzymatic digest of soy. Due to the naturally high concentrations of vitamins and carbohydrates in soy, Phytone Peptone is a nutritious and excellent carbohydrate source.
- Enzymatic digests of soybean meal/flour
- Retains the high vitamin and high carbohydrate content of the soy plant tissue
- It is an excellent plant peptone for the cultivation of fungi and fastidious types of bacteria, such as members of the Clostridium and Neisseria genera
- It has been used in mammalian cell culture applications due to its high carbohydrate content
Customers have successfully used this supplement in CHO-based production of monoclonal antibodies for oncological and rheumatological therapeutics, as well as with many different microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines.
: LUO
- Environmentally Preferable:
Frequently Bought Together