Difco™ Tryptone Peptone, Gibco™ (a part of Thermo Fisher Scientific)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Difco Tryptone Peptone is an animal origin (AO) pancreatic digest of casein. In addition to providing a unique nutritional profile, it has a higher nitrogen content than our other casein products.
- Animal origin, pancreatic digest of casein
- Primary nitrogen source of trypticase soy broth and agar
- Referenced in Official Methods of Analysis of AOAC International
- Supports the growth of many different types of bacteria and fungi
- Recommended for use in media formulations when good growth of fungi and bacteria is required
- Used in the development of animal health vaccines and with human health Botox-like products
Tryptone Peptone supports the growth of many different types of bacteria and fungi, including fastidious organisms and in toxin production. Customers have used this supplement in the development of animal health vaccines, such as Clostridial vaccines, and with human health Botox-like products produced from Clostridium botulinum.
Certifications: LUO
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together