Difco™ Soytone, Gibco™ (a part of Thermo Fisher Scientific)
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Difco Soytone is an enzymatic digest of soy. With high concentrations of vitamins and carbohydrates uniquely present in soy, this supplement offers an alternative carbohydrate profile to yeast supplements for mammalian cell processes, such as CHO.
- High concentrations of vitamins and carbohydrates
- An enzymatic digest of soy
Frequently Bought Together