Bacto™ Casitone, Gibco™ (a part of Thermo Fisher Scientific)
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Bacto Casitone is an animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest.
- Pancreatic digest of casein
- More nutritious hydrolysate, especially for those organisms that prefer peptides to amino acids
- Can be used as a component in microbiological media or in fermentation applications
While this supplement provides a nutritional profile based on casein, the specific enzymatic digest performed for this product results in a particularly high content of amino acids and peptides of varying sizes, making it a more nutritious hydrolysate. Can be used for the production of Escherichia coli based recombinant proteins.
Frequently Bought Together