The refrigerated Eppendorf Centrifuge 5427 R supports your greener lab with hydrocarbon cooling. It's an ideal solution for laboratories with a variety of molecular biology applications or who are in need of high-throughput solutions for 1.5 to 2 ml tubes - all combined in a microcentrifuge with a high throughput/footprint-ratio.
- Cooled with natural refrigerant (R290) to protect samples and the planet
- Soft touch one-finger closure for ergonomic operation
- FastTemp function for quick pre-cooling of the centrifuge, e.g., from ~23 to 4 °C in only 11 minutes
- ECO shut-off can be programmed to engage after 1, 2, 4 or 8 hours of non-use for maximum energy savings and extended compressor life
- Built-in condensation drain to eliminate water accumulation
- Automatic rotor recognition and imbalance detection for maximum operational safety
- No.of.rotors available: Nine rotors
The refrigerated microcentrifuge Centrifuge 5427 R was designed with high-throughput research applications in mind. With its aerosol-tight 48-place rotor it handles even the toughest challenges in a safe and reliable manner to provide the efficiency boost you need. With speeds of up to 25,001×g, this microcentrifuge is fast enough for all your molecular biology applications. All aerosol-tight rotors are equipped with the Eppendorf QuickLock lids for easy rotor lid opening and closing to enable safe centrifugation of hazardous samples. Choose the Centrifuge 5427 R if a 24-place microcentrifuge does not meet your needs regarding tube capacity and if you don't need the extra versatility our cross-over Centrifuge 5430R can offer.
- Catalog No:
- CA76637-332
- CA76637-336
- CA76637-334
- Environmentally Preferable:
- Safer Human and Environmental Health, Low Manufacturing Impact
- Safer Human and Environmental Health, Low Manufacturing Impact
- Safer Human and Environmental Health, Low Manufacturing Impact
- Max. capacity:
- 48×1.5/2.0 ml
- 48×1.5/2.0 ml
- 48×1.5/2.0 ml
- Max. speed:
- 16220 rpm
- 16220 rpm
- 16220 rpm
- Max. RCF:
- 25001×g
- 25001×g
- 25001×g
- Refrigerated:
- Yes
- Yes
- Yes
- Dimensions:
- 31.9×54.0×25.4 cm (12.6×21.3×10")
- 31.9×54.0×25.4 cm (12.6×21.3×10")
- 31.9×54.0×25.4 cm (12.6×21.3×10")
- Weight:
- 30 kg (66.1 lbs.)
- 30 kg (66.1 lbs.)
- 30 kg (66.1 lbs.)
- Environmental certification:
- Renewable Energy Certificate; ISO 14001 (Cert No: 12 104 43612 TMS); EPA SNAP Acceptable refrigerant R290 (Propane).
- Renewable Energy Certificate; ISO 14001 (Cert No: 12 104 43612 TMS); EPA SNAP Acceptable refrigerant R290 (Propane).
- Renewable Energy Certificate; ISO 14001 (Cert No: 12 104 43612 TMS); EPA SNAP Acceptable refrigerant R290 (Propane).